Learn More
Bachem BKT140 Acetate
SDP
Supplier:  Bachem 4073017.1000
Encompass_Preferred
BKT140 Acetate, 4-Fluorobenzoyl-Arg-Arg-2-Nal-Cys-Tyr-Cit-Lys-D-Lys-Pro-Tyr-Arg-Cit-Cys-Arg-NH2, 664334-36-5, C97H144FN33O19S2, Mr 2159.55, 1mg. 4F-benzoyl-TN14003, BL-8040. As described by Peled and coworkers, BKT140 (4F-benzoyl-TN14003) is a biostable, high-affinity chemokine receptor CXCR4 antagonist, which binds CXCR4 with a greater affinity than plerixafor. When combined with G-CSF (granulocyte colony-stimulating factor), BKT140 is a safe and efficient stem cell mobilizer that enabled the collection of a high number of CD34+ cells in 1 and 2 aphaeresis procedures, resulting in successful engraftment.
Catalog No. 50-194-6732