Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AS-Inclisiran sodium | 00-00-0 | 94.3% | C237H291F10N101O134P22S4 | 10MG
SDP

Catalog No. 5000261129
Encompass_Preferred

AS-Inclisiran sodium is an antisense oligonucleotide designed as the complementary strand to inclisiran to modulate PCSK9 expression for lipid-lowering research. Supplied as a white to off-white solid, it is intended for in vitro and preclinical studies.

  • Molecular weight 7699.6 Da.
  • Molecular formula C237H291F10N101O134P22S4.
  • Targets PCSK9 (proprotein convertase subtilisin/kexin type 9).
  • Purity approximately 94.3%.
  • Soluble in water at ≥100 mg/mL.
  • Store at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Appearance white to off-white solid.

Catalog No. 50-002-61129 Supplier Medchemexpress LLC Supplier No. HY14322110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AS-Inclisiran sodium is an antisense oligonucleotide designed as the complementary strand to inclisiran to modulate PCSK9 expression for lipid-lowering research. Supplied as a white to off-white solid, it is intended for in vitro and preclinical studies.

  • Molecular weight 7699.6 Da.
  • Molecular formula C237H291F10N101O134P22S4.
  • Targets PCSK9 (proprotein convertase subtilisin/kexin type 9).
  • Purity approximately 94.3%.
  • Soluble in water at ≥100 mg/mL.
  • Store at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Appearance white to off-white solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.