Learn More
Bachem Cyclo(-Gln-Gly)
SDP
Supplier:  Bachem 4014279.1000
Encompass_Preferred
Cyclo(-Gln-Gly), 52662-00-7, C7H11N3O3, Mr 185.18, 1g. As its linear analog, N-1070, cyclo(Gly-Gln) is a biologically active peptide. The diketopiperazine is capable of reversing the cardiorespiratory depression produced by b-endorphin or morphine.
Catalog No. 50-194-6776