Learn More
As One International Inc AS ONE Glove Box, PMMA, 800 x 500 x 528 mm, 1 ea
Catalog No. NC3929662
Encompass_Preferred
This acrylic glove box is designed for research and production environments requiring gas replacement or controlled atmospheres. Constructed from transparent PMMA, it provides high internal visibility and a spacious working environment measuring 800 x 500 x 528 mm. The unit includes integrated electrical outlets and gas management features to support a range of laboratory applications.
  • Equipped with a pass box for convenient internal access and inspection
  • Constructed from durable transparent material for high visibility
  • Supports gas replacement with two included cocks for φ10 mm tubing
  • Features two AC 100 V electrical outlets for auxiliary equipment
  • Includes a safety valve to release pressure during excessive internal pressure rise
  • Comes with one pair of large gloves with a 0.4 mm thickness
Catalog No. NC3929662 Supplier As One International Inc Supplier No. 1904602
This acrylic glove box is designed for research and production environments requiring gas replacement or controlled atmospheres. Constructed from transparent PMMA, it provides high internal visibility and a spacious working environment measuring 800 x 500 x 528 mm. The unit includes integrated electrical outlets and gas management features to support a range of laboratory applications.
  • Equipped with a pass box for convenient internal access and inspection
  • Constructed from durable transparent material for high visibility
  • Supports gas replacement with two included cocks for φ10 mm tubing
  • Features two AC 100 V electrical outlets for auxiliary equipment
  • Includes a safety valve to release pressure during excessive internal pressure rise
  • Comes with one pair of large gloves with a 0.4 mm thickness