Learn More
Medchemexpress LLC AS-Patisiran sodium | 1 MG
SDP
Catalog No. 5000373330
Encompass_Preferred
AS-Patisiran sodium is an antisense strand of Patisiran. Patisiran is a double-stranded small interfering RNA (siRNA) that targets a sequence within the transthyretin (TTR) messenger RNA. It specifically inhibits hepatic synthesis of mutant and wild-type TTR.
  • Targets a sequence within the transthyretin (TTR) messenger RNA
  • Specifically inhibits hepatic synthesis of mutant and wild-type TTR
  • Can be used for the research of hereditary TTR amyloidosis
Catalog No. 50-003-73330 Supplier Medchemexpress LLC Supplier No. HY1536091MG
AS-Patisiran sodium is an antisense strand of Patisiran. Patisiran is a double-stranded small interfering RNA (siRNA) that targets a sequence within the transthyretin (TTR) messenger RNA. It specifically inhibits hepatic synthesis of mutant and wild-type TTR.
  • Targets a sequence within the transthyretin (TTR) messenger RNA
  • Specifically inhibits hepatic synthesis of mutant and wild-type TTR
  • Can be used for the research of hereditary TTR amyloidosis