Learn More
Perkin Elmer US LLC Stainless Steel Sample Probe with Filtered Tip
Catalog No. 509043813
Encompass_Preferred
The stainless steel sample probe features a screen tip and is used in oils applications. The probe includes the capillary tube fitting, although doesn't include capillary tubing.
Catalog No. 50-904-3813 Supplier Perkin Elmer US LLC Supplier No. N0771529
The stainless steel sample probe features a screen tip and is used in oils applications. The probe includes the capillary tube fitting, although doesn't include capillary tubing.