Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AS057278 | 402-61-9 | 99.76% | 126.11 | 200 MG
SDP

Catalog No. 5000404020
Encompass_Preferred

AS057278 is a potent, selective, orally active, and blood-brain barrier (BBB) penetrant non-peptidic D-amino acid oxidase (DAAO) inhibitor with an IC50 value of 0.91 μM and EC50 of 2.2-3.95 μM. It can normalize phencyclidine (PCP)-induced prepulse inhibition in mice and can be used for researching schizophrenia.

  • Inhibits D-amino acid oxidase (DAAO)
  • Potent, selective, orally active, and BBB penetrant
  • Normalizes phencyclidine (PCP)-induced prepulse inhibition in mice
  • Exhibits good pharmacokinetic effects in male Sprague-Dawley rats
  • Appearance: solid, light yellow to light brown
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-04020 Supplier Medchemexpress LLC Supplier No. HY33009200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AS057278 is a potent, selective, orally active, and blood-brain barrier (BBB) penetrant non-peptidic D-amino acid oxidase (DAAO) inhibitor with an IC50 value of 0.91 μM and EC50 of 2.2-3.95 μM. It can normalize phencyclidine (PCP)-induced prepulse inhibition in mice and can be used for researching schizophrenia.

  • Inhibits D-amino acid oxidase (DAAO)
  • Potent, selective, orally active, and BBB penetrant
  • Normalizes phencyclidine (PCP)-induced prepulse inhibition in mice
  • Exhibits good pharmacokinetic effects in male Sprague-Dawley rats
  • Appearance: solid, light yellow to light brown
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.