Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific AS12A GUARD COLUMN 4MM

Catalog No. NC9959374 Shop All Thermo Scientific Products
Encompass

IonPac AS12A Analytical Column (4 x 200 mm)

Catalog No. NC9959374 Supplier Thermo Scientific Supplier No. 046034
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IonPac AS12A Analytical Column (4 x 200 mm)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.