Learn More
Thermo Scientific AS14A-7 IONPAC COLUMN
Catalog No. NC9314210 Shop All Thermo Scientific Products
Encompass
IonPac AS14A-7 um Analytical Column (4 x 250 mm)
Catalog No. NC9314210 Supplier Thermo Scientific Supplier No. 056904
IonPac AS14A-7 um Analytical Column (4 x 250 mm)