Learn More
Selleck Chemical LLC AS1517499 5mg 919486-40-1
SDP
Supplier:  Selleck Chemical LLC S86855MG
Encompass_Preferred
AS1517499 is a novel and potent STAT6 inhibitor with an IC50 value of 21 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8102