Learn More
Medchemexpress LLC HY-100614 5mg Medchemexpress, AS1517499 CAS:919486-40-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006145MG
Encompass_Preferred
Medchemexpress, HY-100614 5mg AS1517499 CAS:919486-40-1 AS1517499 is a potent STAT6 inhibitor with an IC 50 of 21 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1901