Learn More
Medchemexpress LLC HY-100614 100mg , AS1517499 CAS:919486-40-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY100614100MG
Encompass_Preferred
Medchemexpress, HY-100614 100mg AS1517499 CAS:919486-40-1 Formula:C20H20ClN5O2 IC50: 21 nM (STAT6) Purity:98% AS1517499 is a potent STAT6 inhibitor with IC 50 of 21 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2984