Learn More
Medchemexpress LLC HY-100596 10mg , AS1842856 CAS:836620-48-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10059610MG
Encompass_Preferred
HY-100596 10mg AS1842856 CAS: 836620-48-5 AS1842856, a specific Foxo1 inhibitor (IC50=30 nM), potently suppresses autophagy. AS1842856 only reduces the activity of FoxO1 by binding with it, without affecting its transcription and protein expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9864