Learn More
Medchemexpress LLC AS1842856 | 836620-48-5 | 99.9% | 347.4 g/mol | C18H22FN3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10059650MG
Encompass_Preferred
AS1842856 is a small-molecule Foxo1 inhibitor used in biochemical and cellular research to reduce Foxo1 transcriptional activity and investigate autophagy-related pathways. It is supplied as a solid and as a 10 mM solution in DMSO for assay and cell-treatment use.
  • Potent Foxo1 inhibitor (IC50 ≈ 30 nM).
  • High purity suitable for research applications (99.88%).
  • Molecular formula C18H22FN3O3 and molecular weight ≈ 347.4 g/mol.
  • Provided as a solid and as a 10 mM solution in DMSO for flexibility in workflows.
  • Suitable for biochemical and cell-based assays studying autophagy and Foxo1 signaling.
  • Registered CAS number 836620-48-5 for unambiguous identification.
Catalog No. 50-202-8629