Learn More
TARGETMOL CHEMICALS INC AS1949490 10MG
SDP
Catalog No. 502577386
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS1949490 activated glucose metabolism via up-regulation of GLUT1 gene in L6 myotubes[1][2]. AS1949490 is a potent and selective SHIP-2 (SH2 domain-containing inositol 5′ phosphatase 2) inhibitor with an IC50 of 620 nM. purity: 99%
Catalog No. 50-257-7386 Supplier TARGETMOL CHEMICALS INC Supplier No. T1432710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS1949490 activated glucose metabolism via up-regulation of GLUT1 gene in L6 myotubes[1][2]. AS1949490 is a potent and selective SHIP-2 (SH2 domain-containing inositol 5′ phosphatase 2) inhibitor with an IC50 of 620 nM. purity: 99%