Learn More
Thermo Scientific Dionex IonPac AS22 IC Columns | | 1 EA
Supplier:  Thermo Scientific 064137
Encompass
Separate common inorganic anions in 8 minutes from diverse matrices using an isocratic carbonate/bicarbonate eluent in a high-capacity, high-resolution column. Designed specifically for compliance monitoring of inorganic anions in accordance with US EPA Methods 3000 A. Fast analysis of drinking water, ground water, and wastewater. Multiple Formats Available for Application Flexibility. Solvent compatibility permits easy column cleanup after the analysis of samples with hydrophobic components.
Catalog No. NC0452266