Learn More
Medchemexpress LLC AS2717638 100mg | 2148339-28-8 | 100 MG
SDP
Catalog No. 5000229788
Encompass_Preferred
AS2717638 is a brain-penetrant, orally active lysophosphatidic acid receptor 5 (LPA5) antagonist used in research on pain and neuroinflammation. It demonstrates high potency for human LPA5 and is supplied with research-grade purity.
  • Brain-penetrant and orally active.
  • High potency for hLPA5 (IC50 38 nM).
  • Molecular formula C25H25N3O5; molecular weight 447.5 g/mol.
  • Supplied at high purity, typically ≥98%.
  • Intended for research use only, not for human therapeutic use.
Catalog No. 50-002-29788 Supplier Medchemexpress LLC Supplier No. HY114379100MG
AS2717638 is a brain-penetrant, orally active lysophosphatidic acid receptor 5 (LPA5) antagonist used in research on pain and neuroinflammation. It demonstrates high potency for human LPA5 and is supplied with research-grade purity.
  • Brain-penetrant and orally active.
  • High potency for hLPA5 (IC50 38 nM).
  • Molecular formula C25H25N3O5; molecular weight 447.5 g/mol.
  • Supplied at high purity, typically ≥98%.
  • Intended for research use only, not for human therapeutic use.