Learn More
Medchemexpress LLC AS2717638 | 25MG
SDP
Supplier:  Medchemexpress LLC HY11437925MG
Encompass_Preferred
AS2717638 is a highly selective brain-penetrant and orally active lysophosphatidic acid receptor 5 (LPA5) antagonist with an IC50 value of 38 nM AS2717638 is highly selective and shows no significant antagonistic activity against other LPA receptors (LPA1 LPA2 and LPA3) AS2717638 can be used in the research of pain and neuroinflammation-related diseases[1][2]
Catalog No. 50-002-28848