Learn More
Medchemexpress LLC AS2717638 50mg | 2148339-28-8 | 50 MG
SDP
Catalog No. 5000229848
Encompass_Preferred
AS-2717638 is a selective, brain-penetrant, orally active lysophosphatidic acid receptor 5 (LPA5) antagonist used in pharmacology and neuroscience research to probe LPA5-mediated signaling and pain models.
  • Selective LPA5 antagonist (IC50 = 0.038 μM) for receptor pharmacology studies.
  • Brain-penetrant and orally active for in vivo models.
  • High purity suitable for biological assays.
  • Stable when stored at -20°C with a long shelf life.
  • Available in small-scale quantities for preclinical research.
Catalog No. 50-002-29848 Supplier Medchemexpress LLC Supplier No. HY11437950MG
AS-2717638 is a selective, brain-penetrant, orally active lysophosphatidic acid receptor 5 (LPA5) antagonist used in pharmacology and neuroscience research to probe LPA5-mediated signaling and pain models.
  • Selective LPA5 antagonist (IC50 = 0.038 μM) for receptor pharmacology studies.
  • Brain-penetrant and orally active for in vivo models.
  • High purity suitable for biological assays.
  • Stable when stored at -20°C with a long shelf life.
  • Available in small-scale quantities for preclinical research.