Learn More
TARGETMOL CHEMICALS INC AS2717638 5MG
SDP
Catalog No. 502581730
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS2717638 is an orally active and selective lysophosphatidic acid receptor 5 (LPA5) antagonist (IC50 38 nM for hLPA5). It also significantly improves PGF2(alpha)- PGE2- and AMPA-induced allodynia. purity: 98%
Catalog No. 50-258-1730 Supplier TARGETMOL CHEMICALS INC Supplier No. T103815MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS2717638 is an orally active and selective lysophosphatidic acid receptor 5 (LPA5) antagonist (IC50 38 nM for hLPA5). It also significantly improves PGF2(alpha)- PGE2- and AMPA-induced allodynia. purity: 98%