Learn More
Medchemexpress LLC HY-114379 5mg Medchemexpress, AS2717638 CAS:2148339-28-8 Purity:>98%
SDP
Catalog No. 501932484
Encompass_Preferred
Medchemexpress, HY-114379 5mg AS2717638 CAS:2148339-28-8 AS2717638 is an oral active and selective lysophosphatidic acid receptor 5 (LPA5) antagonist, with an IC 50 of 38 nM for hLPA5. AS2717638 also significantly improves PGE 2 -, PGF 2α -, and AMPA-induced allodynia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2484 Supplier Medchemexpress LLC Supplier No. HY1143795MG
Medchemexpress, HY-114379 5mg AS2717638 CAS:2148339-28-8 AS2717638 is an oral active and selective lysophosphatidic acid receptor 5 (LPA5) antagonist, with an IC 50 of 38 nM for hLPA5. AS2717638 also significantly improves PGE 2 -, PGF 2α -, and AMPA-induced allodynia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.