Learn More
Selleck Chemical LLC AS2863619 S8903-100MG
SDP
Catalog No. 5000024216
Encompass_Preferred
AS2863619 is a small-molecule cyclin-dependent kinase CDK8/19 inhibitor with IC50 of 0 6099 nM and 4 277 nM respectively AS2863619 is a potent Foxp3 inducer in Tconv cells
Catalog No. 50-000-24216 Supplier Selleck Chemical LLC Supplier No. S8903100MG
AS2863619 is a small-molecule cyclin-dependent kinase CDK8/19 inhibitor with IC50 of 0 6099 nM and 4 277 nM respectively AS2863619 is a potent Foxp3 inducer in Tconv cells