Learn More
Selleck Chemical LLC AS2863619 2mg
SDP
Supplier:  Selleck Chemical LLC S89032MG
Encompass_Preferred
AS2863619 is a small-molecule cyclin-dependent kinase CDK8/19 inhibitor with IC50 of 0.6099 nM and 4.277 nM, respectively. AS2863619 is a potent Foxp3 inducer in Tconv cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3426