Learn More
TARGETMOL CHEMICALS INC AS2863619 5MG
SDP
Catalog No. 502579083
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS2863619 is an orally active inhibitor of cyclin-dependent kinase 8 (CDK8) and CDK19 (IC50s of 0.61 nM and 4.28 nM respectively). purity: 100%
Catalog No. 50-257-9083 Supplier TARGETMOL CHEMICALS INC Supplier No. T83785MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS2863619 is an orally active inhibitor of cyclin-dependent kinase 8 (CDK8) and CDK19 (IC50s of 0.61 nM and 4.28 nM respectively). purity: 100%