Learn More
Medchemexpress LLC HY-126675A 5mg Medchemexpress, AS2863619 CAS:2241300-51-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY126675A5MG
Encompass_Preferred
Medchemexpress, HY-126675A 5mg AS2863619 CAS:2241300-51-4 AS2863619 enables conversion of antigen-specific effector/memory T cells into Foxp3+ regulatory T (Treg) cells for the treatment of various immunological diseases. AS2863619 is a potent, orally active cyclin-dependent kinase 8 (CDK8) and CDK19 inhibitor with IC50s of 0.61 nM and 4.28 nM, respectively. STAT5 activation enhanced by AS2863619 inhibition of CDK8/19, which consequently activates the Foxp3 gene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9736