Learn More
Aviva Systems Biology Corp AS3MT Antibody - middle region (ARP48862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AS3MT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AS3MT. Alternate names: CYT19, RP11-753C18.6ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-383