Learn More
Thermo Scientific AS4A COLUMN 4MMAS4A COLUMN 4
Supplier:  Thermo Scientific 043174
Encompass
IonPac AS4A-SC Analytical Column, (4 x 250 mm), Maximum Operating Pressure:4000 psi, Mobile Phase Compatibility:pH 0-14; 0-100% HPLC Solvents, Capacity:20 microeq, Bead Diameter: 13 micrometer, Crosslinking (%DVB): 55%, Functional Group:Alk
Catalog No. NC9814607