Learn More
Marco Rubber AS568-018 O-rings FFKM, Markez® Z1028 75 Durometer, Black, FFKM Perfluoroelastomer
SDP
Catalog No. NC3584549
Encompass
Markez® Z1028-018 is a 75 durometer, black, FFKM Perfluoroelastomer o-ring. This product is a USA AS568-018 size and has a cross section of 1.78mm (0.070in) and an inside diameter of 18.77mm (0.739in). It is made from the compound Markez® Z1028. See compound features below
Catalog No. NC3584549 Supplier Marco Rubber Supplier No. Z1028018
Markez® Z1028-018 is a 75 durometer, black, FFKM Perfluoroelastomer o-ring. This product is a USA AS568-018 size and has a cross section of 1.78mm (0.070in) and an inside diameter of 18.77mm (0.739in). It is made from the compound Markez® Z1028. See compound features below