Learn More
Enzo Life Sciences AS601245 (1mg). CAS: 345987-15-7
SDP
Supplier:  Enzo Life Sciences ALX270443M001
Encompass_Preferred
Potent and cell permeable ATP competitive JNK inhibitor (hJNK1: IC50=150nM, hJNK2: IC50=220nM and hJNK3: IC50=70 nM). Displays anti-inflammatory properties and has been shown to reduce TNF-α plasma levels induced by LPS in mice. Alternative name: 1,3-Benzothiazol-2-yl-(2-{[2-(3-pyridinyl)ethyl]amino}-4-pyrimidinyl) acetonitrile, JNK Inhibitor V. Purity: ≥95% (HPLC). Appearance: White to yellow solid. Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8565