Learn More
Medchemexpress LLC HY-11010 50mg , AS601245 CAS:345987-15-7 Purity:98%
SDP
Catalog No. 501493832
Encompass_Preferred
Medchemexpress, HY-11010 50mg AS601245 CAS:345987-15-7 Formula:C20H16N6S IC50: 70 nM (hJNK3), 150 nM (hJNK1), 220 nM (hJNK2) Purity:98% AS601245 is a JNK Inhibitor with IC 50 s of 150, 220, and 70 nM for three JNK human isoforms ( hJNK1 , hJNK2 , and hJNK3 ), respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3832 Supplier Medchemexpress LLC Supplier No. HY1101050MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-11010 50mg AS601245 CAS:345987-15-7 Formula:C20H16N6S IC50: 70 nM (hJNK3), 150 nM (hJNK1), 220 nM (hJNK2) Purity:98% AS601245 is a JNK Inhibitor with IC 50 s of 150, 220, and 70 nM for three JNK human isoforms ( hJNK1 , hJNK2 , and hJNK3 ), respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.