Learn More
Medchemexpress LLC HY-11010 5mg Medchemexpress, AS601245 CAS:345987-15-7 Purity:>98%
SDP
Catalog No. 501968745
Encompass_Preferred
Medchemexpress, HY-11010 5mg AS601245 CAS:345987-15-7 AS601245 is a cell-permeable JNK Inhibitor with IC50s of 150, 220, and 70 nM for three JNK human isoforms (hJNK1, hJNK2, and hJNK3), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8745 Supplier Medchemexpress LLC Supplier No. HY110105MG
Medchemexpress, HY-11010 5mg AS601245 CAS:345987-15-7 AS601245 is a cell-permeable JNK Inhibitor with IC50s of 150, 220, and 70 nM for three JNK human isoforms (hJNK1, hJNK2, and hJNK3), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.