Learn More
Medchemexpress LLC HY-100744 10mg Medchemexpress, AS8351 CAS:796-42-9 Purity:>98%
SDP
Catalog No. 501931936
Encompass_Preferred
Medchemexpress, HY-100744 10mg AS8351 CAS:796-42-9 AS8351 is a KDM5B inhibitor, which can induce and sustain active chromatin marks to facilitate the induction of cardiomyocyte-like cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1936 Supplier Medchemexpress LLC Supplier No. HY10074410MG
Medchemexpress, HY-100744 10mg AS8351 CAS:796-42-9 AS8351 is a KDM5B inhibitor, which can induce and sustain active chromatin marks to facilitate the induction of cardiomyocyte-like cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.