Learn More
TARGETMOL CHEMICALS INC AS8351 25MG
SDP
Catalog No. 502576809
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS8351 (NSC51355) is a histone demethylase inhibitor. It has been used in combination with CHIR99021 A 83-01 BIX01294 SC-1 Y-27632 OAC2 SU 16f and JNJ-10198409 to induce reprogramming of human fetal lung fibroblasts into functional cardiomyocytes. purity: 99%
Catalog No. 50-257-6809 Supplier TARGETMOL CHEMICALS INC Supplier No. T410025MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS8351 (NSC51355) is a histone demethylase inhibitor. It has been used in combination with CHIR99021 A 83-01 BIX01294 SC-1 Y-27632 OAC2 SU 16f and JNJ-10198409 to induce reprogramming of human fetal lung fibroblasts into functional cardiomyocytes. purity: 99%