Learn More
Hach Company AS950 All-Weather Refrigerated Sampler, 2 Sensor Ports, 115 V, Discrete (4 x 10 L) PE Bottle Kit
Catalog No. NC2809108 Shop All Hach Company Products
Encompass
The AS950 All-Weather Refrigerated Sampler is a durable outdoor automated water sampler designed for reliable sample preservation and intuitive operation. It provides refrigerated sample storage, dual sensor ports for process monitoring, a high-speed peristaltic pump, and a kit of four 10 L polyethylene bottles.
  • Large full-color display with single-screen programming for intuitive operation.
  • USB support for easy data transfer and program copying between samplers.
  • Program status screen shows alarms, missed samples, and progress for quick troubleshooting.
  • All-weather base resists corrosion, rodents, and standing water for outdoor use.
  • Air-sensing thermostat maintains sample temperature at 4 (±0.8) °C for preservation.
  • High-speed dual-roller peristaltic pump with spring-mounted rollers for long tubing life.
  • Includes four 10 L polyethylene sample bottles and vinyl tubing kit.
Catalog No. NC2809108 Supplier Hach Company Supplier No. ASA.CSXX1X31XX
Add to Cart
Add to Cart
Request a Quote
The AS950 All-Weather Refrigerated Sampler is a durable outdoor automated water sampler designed for reliable sample preservation and intuitive operation. It provides refrigerated sample storage, dual sensor ports for process monitoring, a high-speed peristaltic pump, and a kit of four 10 L polyethylene bottles.
  • Large full-color display with single-screen programming for intuitive operation.
  • USB support for easy data transfer and program copying between samplers.
  • Program status screen shows alarms, missed samples, and progress for quick troubleshooting.
  • All-weather base resists corrosion, rodents, and standing water for outdoor use.
  • Air-sensing thermostat maintains sample temperature at 4 (±0.8) °C for preservation.
  • High-speed dual-roller peristaltic pump with spring-mounted rollers for long tubing life.
  • Includes four 10 L polyethylene sample bottles and vinyl tubing kit.