Learn More
Hach Company AS950 Portable Compact Sampler
Supplier:  Hach Company ASP.CXXXC261XX
Encompass
AS950 Portable Compact Sampler-115 V-Discrete (24x) 575 mL PE Bottle Kit-US plugIncludes1 ea AS950 controller on compact portable base1 ea 115V power supply (8754500US)1 ea Bottle kit (PC240575) includes 24 - 575mL polyethylene bottles w/caps (1369) retainer (1422) and distributor arm (8580)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)(Battery charger 8753500US sold separately)
Catalog No. 50-002-07722
Add to Cart
Request a Quote