Learn More
Hach Company AS950 Portable Std. Sampler
Catalog No. 5000207546 Shop All Hach Company Products
Encompass
AS950 Portable Standard Sampler-2 Sensor Port Options-12 V-Composite 10 L PE Bottle-Submerged Area Velocity and digital pH SensorIncludes1 ea AS950 controller with sensor ports on standard portable base1 ea 12V battery (8754400)1 ea Bottle kit (PS010030) includes 2 5 gallon polyethylene bottle w/cap (1918)-container support (1502) and full bottle shut-off (8996)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea Non oil-filled 0-10 range submerged area velocity sensor with 30 cable (77065-030)1 ea AV9000S Area Velocity Analyzer (9504600)1 ea AV9000S mounting hardware (9506900)1 ea digital pH sensor (DPD2P1)1 ea pH adapter cable (9501200)(Battery charger 8753500US sold separately)
Catalog No. 50-002-07546 Supplier Hach Company Supplier No. ASP.CSXXS12113
Add to Cart
Add to Cart
Request a Quote
AS950 Portable Standard Sampler-2 Sensor Port Options-12 V-Composite 10 L PE Bottle-Submerged Area Velocity and digital pH SensorIncludes1 ea AS950 controller with sensor ports on standard portable base1 ea 12V battery (8754400)1 ea Bottle kit (PS010030) includes 2 5 gallon polyethylene bottle w/cap (1918)-container support (1502) and full bottle shut-off (8996)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea Non oil-filled 0-10 range submerged area velocity sensor with 30 cable (77065-030)1 ea AV9000S Area Velocity Analyzer (9504600)1 ea AV9000S mounting hardware (9506900)1 ea digital pH sensor (DPD2P1)1 ea pH adapter cable (9501200)(Battery charger 8753500US sold separately)