Learn More
Hach Company AS950 Portable Std. Sampler
Supplier:  Hach Company ASP.CSXXS121XX
Encompass
AS950 Portable Standard Sampler-2 Sensor Port Options-12 V-Composite 2 5 Gallon PE BottleIncludes1 ea AS950 controller with sensor ports on standard portable base1 ea 12V battery (8754400)1 ea Bottle kit (PS010030) includes 2 5 gallon polyethylene bottle w/cap (1918)-container support (1502) and full bottle shut-off (8996)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)(Battery charger 8753500US sold separately)
Catalog No. 50-002-08047
Add to Cart
Request a Quote