Learn More
Hach Company AS950 Portable Std. Sampler
Supplier:  Hach Company ASP.CXXXS241XX
Encompass
AS950 Portable Standard Sampler-115 V-Discrete (24x) 1 L Bottle Kit-US plugIncludes1 ea AS950 controller on standard portable base1 ea 100-120V-3 pin to sampler-US plug1 ea Bottle kit (PS241000) includes 24x1 liter polyethylene bottles w/caps (737) retainer (1422) and distributor arm (8582)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)(Battery charger 8753500US sold separately)
Catalog No. 50-002-07780
Add to Cart
Request a Quote