Learn More
Aviva Systems Biology Corp ASAH1 Antibody - N-terminal region (ARP57760_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASAH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ASAH1. Alternate names: AC, ASAH, FLJ21558, FLJ22079, PHP, PHP32, ACDaseApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62147