Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ASAH1 Human His-SU 100ug
SDP

Catalog No. 5000196386
Encompass_Preferred

Recombinant human ASAH1 (acid ceramidase 1) supplied as an N-SUMO/6xHis-tagged protein, expressed in Escherichia coli and provided lyophilized for research use (100 μg). It hydrolyzes ceramide into sphingosine and fatty acids at acidic pH and includes formulation and storage guidance to preserve stability.

  • Recombinant human enzyme supplied as N-SUMO and N-6xHis tagged protein.
  • Expressed in Escherichia coli and purified to >90% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 58.7 kDa.
  • Lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Unit size 100 μg; do not reconstitute to a concentration below 100 μg/mL.
  • Enzyme activity testing is in progress; activity is not guaranteed.
  • Store lyophilized material at -20°C; after reconstitution keep at 4°C for up to 1 week or at -20°C for long-term storage and avoid repeated freeze-thaw.

Catalog No. 50-001-96386 Supplier Medchemexpress LLC Supplier No. HYP71541100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ASAH1 (acid ceramidase 1) supplied as an N-SUMO/6xHis-tagged protein, expressed in Escherichia coli and provided lyophilized for research use (100 μg). It hydrolyzes ceramide into sphingosine and fatty acids at acidic pH and includes formulation and storage guidance to preserve stability.

  • Recombinant human enzyme supplied as N-SUMO and N-6xHis tagged protein.
  • Expressed in Escherichia coli and purified to >90% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 58.7 kDa.
  • Lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Unit size 100 μg; do not reconstitute to a concentration below 100 μg/mL.
  • Enzyme activity testing is in progress; activity is not guaranteed.
  • Store lyophilized material at -20°C; after reconstitution keep at 4°C for up to 1 week or at -20°C for long-term storage and avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.