Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ASAH1 Human His-SU 10ug
SDP

Catalog No. 5000196750
Encompass_Preferred

Recombinant human ASAH1 (acid ceramidase) is supplied as an N-terminal SUMO-6xHis fusion expressed in E. coli and provided as a lyophilized powder. The product is sequence-verified and characterized for purity and stability, and is intended for biochemical assays, enzyme characterization, and antibody validation.

  • Recombinant human acid ceramidase expressed in E. coli.
  • N-terminal SUMO and 6xHis fusion tag for purification and detection.
  • Sequence corresponds to human ASAH1 residues Q22-W395.
  • Approximate molecular weight of 58.7 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized formulation in Tris-EDTA with trehalose for enhanced stability.
  • Recommend reconstitution to ≥100 μg/mL in ddH2O and aliquot storage at -20°C or -80°C.

Catalog No. 50-001-96750 Supplier Medchemexpress LLC Supplier No. HYP7154110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ASAH1 (acid ceramidase) is supplied as an N-terminal SUMO-6xHis fusion expressed in E. coli and provided as a lyophilized powder. The product is sequence-verified and characterized for purity and stability, and is intended for biochemical assays, enzyme characterization, and antibody validation.

  • Recombinant human acid ceramidase expressed in E. coli.
  • N-terminal SUMO and 6xHis fusion tag for purification and detection.
  • Sequence corresponds to human ASAH1 residues Q22-W395.
  • Approximate molecular weight of 58.7 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized formulation in Tris-EDTA with trehalose for enhanced stability.
  • Recommend reconstitution to ≥100 μg/mL in ddH2O and aliquot storage at -20°C or -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.