Learn More
Medchemexpress LLC ASAH1 Human His-SU 20ug
SDP
Supplier:  Medchemexpress LLC HYP7154120UG
Encompass_Preferred
Recombinant human acid ceramidase (ASAH1) is provided as an N-terminal SUMO-6xHis tagged protein expressed in E. coli and supplied lyophilized from a Tris-EDTA-trehalose buffer. The material is intended for biochemical and sphingolipid metabolism research, offered in microgram-scale pack sizes, and carries storage and handling guidance to preserve integrity. Enzymatic activity testing is ongoing and not guaranteed by the manufacturer.
  • Expressed in E. coli for recombinant production.
  • N-terminal SUMO and 6xHis tag for purification and detection.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized in stabilizing buffer for extended shelf life.
  • Available in multiple microgram pack sizes for small-scale studies.
  • Store at -20°C and aliquot to avoid freeze-thaw cycles.
Catalog No. 50-001-95899