Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 300 x 7.5 mm

Catalog No. 502395657 Shop All Phenomenex Products
Encompass

Asahipak 5 m GF-310 HQ CH0-2905

Catalog No. 50-239-5657 Supplier Phenomenex Inc Supplier No. CH02905
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Asahipak 5 m GF-310 HQ CH0-2905

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.