Learn More
Aviva Systems Biology Corp ASAHL Antibody - middle region (ARP44939_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44939_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASAHL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ASAHL. Alternate names: NAAA, PLT, ASAHLApplicationWB, IHC. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58941