Learn More
Sino Biological ASAM / CLMP Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10794MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ASAM extracellular domain (rh ASAM; Catalog#10794-H08H; NP_079045.1; Met 1-Met 233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7638