Learn More
Waters Corp Waters ASAP Accessory, SYNAPT G2-Si, Xevo G2 Tof, G2 QTof
Catalog No. 50179204 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ASAP accessory enables rapid, direct analysis of volatile and semi-volatile samples for mass spectrometry. It utilizes atmospheric pressure ionization for sensitive analysis of low-polarity compounds and complex mixtures without prior sample preparation.
  • Rapid, direct analysis of volatile and semi-volatile solid and liquid samples.
  • Atmospheric pressure ionization: heated nitrogen desolvation and corona discharge.
  • Sensitive analysis of low-polarity compounds and complex mixtures; no prior sample preparation.
  • Probe body, removable sample insertion assembly, corona pin, glass capillaries included.
  • Fits standard API sources, replacing ESI or APCI probe assemblies.
  • Fixed/removable sections for quick sample introduction, maintaining stable source conditions.
  • Compatible with mass spectrometry instrument series: ACQUITY SQD/TQD, Xevo TQ MS.
Catalog No. 50-179-204 Supplier Waters Corp Supplier No. 176002472
Add to Cart
Add to Cart
Request a Quote
This ASAP accessory enables rapid, direct analysis of volatile and semi-volatile samples for mass spectrometry. It utilizes atmospheric pressure ionization for sensitive analysis of low-polarity compounds and complex mixtures without prior sample preparation.
  • Rapid, direct analysis of volatile and semi-volatile solid and liquid samples.
  • Atmospheric pressure ionization: heated nitrogen desolvation and corona discharge.
  • Sensitive analysis of low-polarity compounds and complex mixtures; no prior sample preparation.
  • Probe body, removable sample insertion assembly, corona pin, glass capillaries included.
  • Fits standard API sources, replacing ESI or APCI probe assemblies.
  • Fixed/removable sections for quick sample introduction, maintaining stable source conditions.
  • Compatible with mass spectrometry instrument series: ACQUITY SQD/TQD, Xevo TQ MS.