Learn More
Aviva Systems Biology Corp ASAP3 Antibody - N-terminal region (ARP57013_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57013_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDEFL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ASAP3. Alternate names: ACAP4, CENTB6, FLJ20199, UPLC1, DDEFL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-54033