Learn More
TARGETMOL CHEMICALS INC ASAPIPRANT 5MG
SDP
Catalog No. 502579778
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Asapiprant (S-555739) is a potent and selective DP1 receptor antagonist (Ki 0.44 nM). It exhibited high affinity and selectivity for the DP1 receptor. purity: 99%
Catalog No. 50-257-9778 Supplier TARGETMOL CHEMICALS INC Supplier No. T53865MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Asapiprant (S-555739) is a potent and selective DP1 receptor antagonist (Ki 0.44 nM). It exhibited high affinity and selectivity for the DP1 receptor. purity: 99%