Learn More
Selleck Chemical LLC Asaraldehyde 10mg 4460-86-0 Asaronaldehyde
SDP
Supplier:  Selleck Chemical LLC S253110MG
Encompass_Preferred
Asaraldehyde (Asaronaldehyde) is a natural COX-2 inhibitor, exhibiting 17-fold selectivity over COX-1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-584-8