Learn More
Cayman Chemical Asarone 100mg
SDP
Catalog No. 502951991
Encompass_Preferred
Antgen purfed eggshell protens (Zrp and small amounts of zona pellucda protens) from Atlantc salmon (S salar) Host mouse clone MN2B4 CrossReactvty ( ) Atlantc salmon (S salar) (reacts prmarly wth B monomer) ranbow trout (O mykss) and Arctc char (S alpnus) Zrp Applcaton(s) WB and ELSA sotype gG1K
Catalog No. 50-295-1991 Supplier Cayman Chemical Supplier No. 11682 100MG
Antgen purfed eggshell protens (Zrp and small amounts of zona pellucda protens) from Atlantc salmon (S salar) Host mouse clone MN2B4 CrossReactvty ( ) Atlantc salmon (S salar) (reacts prmarly wth B monomer) ranbow trout (O mykss) and Arctc char (S alpnus) Zrp Applcaton(s) WB and ELSA sotype gG1K